Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APIP, Mouse anti-Human, Clone: 19F424, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280579
Description
APIP Monoclonal antibody specifically detects APIP in Human Samples. It is validated for Western Blot.Specifications
APIP | |
19F424 | |
Unconjugated | |
PBS and 0.2% Gelatin | |
APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase | |
Mouse | |
IgG1 Kappa | |
0.1 mg | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Cross reacts with Human. |
Western Blot | |
0.5 mg/mL | |
Western Blot 1-2 ug/ml | |
Purified | |
APIP | |
The antibody was developed against a full-length His-tagged recombinant APIP protein. | |
Protein G purified | |
RUO | |
Primary | |
51074 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title