Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APJ/Apelin receptor Mouse anti-Human, Alexa Fluor 405, Clone: 72133R, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8561V100UG
This item is not returnable.
View return policy
Description
APJ/Apelin receptor Monoclonal specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry.Specifications
APJ/Apelin receptor | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
AGTRL1, AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, APLNR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11, HG11 orphan receptor, MGC45246 | |
Mouse myeloma cell line NS0-derived recombinant human APJ Met1-Asp380 Accession # P35414 | |
RUO | |
187 | |
Human | |
IgG1 |
Flow Cytometry | |
72133R | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P35414 | |
Mouse | |
100 μg | |
Primary | |
Detects human APJ in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)