Learn More
APJ/Apelin receptor Mouse anti-Human, Alexa Fluor 594, Clone: 72133R, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8561T100UG
Catalog No. FAB8561T100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
APJ/Apelin receptor
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
AGTRL1, AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, APLNR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11, HG11 orphan receptor, MGC45246
Mouse myeloma cell line NS0-derived recombinant human APJ Met1-Asp380 Accession # P35414
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human APJ in direct ELISAs.
Flow Cytometry
72133R
Flow Cytometry 0.25-1 ug/10^6 cells
P35414
Mouse
IgG1
RUO
Primary
187
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)