Learn More
APOBEC2 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29962
Catalog No. 89-033-250
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human APOBEC2.
  • High specificity
  • High reactivity
Specifications
Specifications
APOBEC2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_006789
ARCD1|ARP1
Rabbit
Protein A purification
RUO
Primary
10930
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human APOBEC2.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
APOBEC2
A synthetic peptide corresponding to N-terminus of human APOBEC2.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)