Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOBEC2 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $534.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen APOBEC2
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3016 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39986
10 μL
Each for $135.30
Add to cart
 
50-199-3015 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39985
100 μL
Each for $534.00
Add to cart
 
Description

Description

APOBEC2 belongs to the cytidine and deoxycytidylate deaminase family. It is probable C to U editing enzyme whose physiological substrate is not yet known. It does not display detectable apoB mRNA editing and has a low intrinsic cytidine deaminase activity.
Specifications

Specifications

Polyclonal
Primary
APOBEC2
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit