Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOBEC3A Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP30450020UL
Description
APOBEC3A Polyclonal antibody specifically detects APOBEC3A in Human samples. It is validated for Western BlotSpecifications
APOBEC3A | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
20 μL | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Western Blot | |
PBS with 50% glycerol, pH7.3. | |
apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A, ARP3, bK150C2.1, EC 3.5.4, EC 3.5.4.-, phorbolin 1, Phorbolin-1, PHRBNprobable DNA dC->dU-editing enzyme APOBEC-3A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human APOBEC3A (NP_663745.1). | |
Affinity purified | |
RUO | |
Primary | |
200315 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title