Learn More
BioVision IncSupplier Diversity Partner ApoBrdU-IHC DNA Fragmentation Assay Kit: Apo-BrdU-IHCTM In Situ DNA Fragmentation Assay Kit: Two-Color TUNEL Assay for Labeling DNA breaks to Detect Apoptotic Cells by Immunohistochemistry. Includes both Positive/Negative Control Slides. 50 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K40350
Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells. BioVision’s Apo-BrdU-IHCTM Kit is a two-color TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for labeling DNA breaks to detect apoptotic cells by immunohistochemistry. The kit contains positive/negative control slides for assessing reagent performance; reaction and blocking buffers for processing individual steps in the assay; proteinase K; terminal deoxynucleotidyl tranferase enzyme (TdT), bromodeoxyuridine triphosphate (Br-dUTP), biotin labeled antiBrdU antibody for labeling DNA breaks, horseradish peroxidase streptavidin conjugate, DAB, H₂O₂/Urea tablets for color generation and methyl green solution for counter staining the cells. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9737