Learn More
Abnova™ APOC1 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L018T6
Catalog No. 89-935-340
Add to cart
Description
Description
Specifications
Specifications
HEK293
BC009698
Theoretical M.W. 9kDa
Transient overexpression cell lysate was tested with anti-APOC1 antibody (Mfr. No. H00000341-M01) by Western Blot.
In modified RIPA Lysis Buffer.
2 mg/mL
APOC1 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-APOC1 full length
100 μg
Human
Immunoprecipitation, Western Blot