Learn More
Aviva Systems Biology Corp APOE ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP54283_P050
This is a rabbit polyclonal antibody against APOE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOE. Alternate names: AD2, MGC1571, apoprotein, LPG, LDLCQ5ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-53735