Learn More
Abnova™ APOL4 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00080832T02
Catalog No. 89-935-380
Add to cart
Description
Description
Specifications
Specifications
293T
ENST00000328429
pCMV-APOL4 full-length
100 μL
Human
APOL4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 11.3kDa
Transient overexpression cell lysate was tested with anti-APOL4 antibody (Mfr. No. H00080832-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot