Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein L4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549586

Catalog No. PA549586

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May play a role in lipid exchange and transport throughout the body. May participate in reverse cholesterol transport from peripheral cells to the liver.
Specifications

Specifications

Apolipoprotein L4
Polyclonal
PBS with 0.09% sodium azide
Q9BPW4
Apolipoprotein L4, Apolipoprotein L-IV, ApoL-IV, APOL4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80832
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
APOL4
Liquid
APOL4
KLH conjugated synthetic peptide between 258-290 amino acids from the Central region of human APOL4.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit