Learn More
APOLD1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 244551AP
Catalog No. 50-173-4368
Add to cart
Description
Description
May be involved in angiogenesis. May play a role in activity-dependent changes of brain vasculature. May affect blood-brain permeability.
Specifications
Specifications
APOLD1
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96LR9
APOLD1, VERGE
Rabbit
IgG
150 μL
-20°C
Primary
81575
Western Blot
Unconjugated
APOLD1
Liquid
APOLD1
APOLD1 Fusion Protein Ag19811
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human