Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Apolipoprotein A-I/ApoA1 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

$293.00 - $629.00

Specifications

Target Apolipoprotein A-I/ApoA1
Conjugate HRP
Target Species Human
Gene ID (Entrez) 335
Gene Symbol APOA1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010051
View Documents
Novus Biologicals™
NBP27950615PLATES
15 Plates
Each for $629.00
Add to cart
 
NB010052
View Documents
Novus Biologicals™
NBP2795065PLATES
5 Plates
Each for $293.00
Add to cart
 
Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human Apolipoprotein A-I/ApoA1.
Specifications

Specifications

Apolipoprotein A-I/ApoA1
Human
APOA1
HRP
335
Rabbit
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit