Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Rabbit anti-Human, Clone: 2083D, Novus Biologicals™
Rabbit Monoclonal Antibody
$206.00 - $493.00
Specifications
Antigen | Apolipoprotein A-I/ApoA1 |
---|---|
Clone | 2083D |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF |
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
Apolipoprotein A-I/ApoA1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
PBS with No Preservative | |
335 | |
rhApoA1 aa25-267 | |
Primary | |
Store at -20C in the dark. Avoid freeze-thaw cycles. |
2083D | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
APOA1 | |
IgG | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title