Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A4 Monoclonal Antibody (CL0468)
Supplier: Thermo Scientific MA534984
Description

Specifications
Apolipoprotein A4 | |
Monoclonal | |
Unconjugated | |
APOA4 | |
APOA4; Apoa-4; APOAIV; apo-AIV; ApoA-IV; Apolipoprotein A4; apolipoprotein AIV; apolipoprotein A-IV; MGC142154; MGC142156 | |
Mouse | |
Protein A | |
RUO | |
337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
CL0468 | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P06727 | |
APOA4 | |
Recombinant Protein corresonding to Human Apolipoprotein A4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction