Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A5, Mouse anti-Human, Clone: 1G5G9, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280580
Description
Apolipoprotein A5 Monoclonal antibody specifically detects Apolipoprotein A5 in Human Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ICC/IF (Negative), CyTOF.Specifications
Apolipoprotein A5 | |
1G5G9 | |
Unconjugated | |
PBS (pH 7.4) | |
apo-AV, APOAV, APOA-V, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
Primary | |
116519 | |
Human |
CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.9 mg/mL | |
Western Blot 0.5 ug/ml, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, ICC/IF (Negative), CyTOF-ready | |
Purified | |
APOA5 | |
Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788] | |
Protein G purified | |
RUO | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title