Learn More
Apolipoprotein A5 Mouse anti-Human, Hamster, Alexa Fluor 700, Clone: 1G5G9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB400139AF700
Catalog No. NBU139AF700
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Apolipoprotein A5
1G5G9
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Unpurified
APOA5
Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
Unpurified
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
116519
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
apo-AV, APOAV, APOA-V, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Hamster