Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein B/ApoB Mouse anti-Human, Alexa Fluor™ 594, Clone: 7B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223519AF594

Catalog No. NP22319A94


Add to cart

Description

Description

Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Apolipoprotein B/ApoB
7B8
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Conjugated, Purified
APOB
Human LDL
Protein A or G purified
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
338.0
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone 02 reacts with apo B from human serum/plasma, when used in sandwich ELISA in combination with a polyclonal antibody against apo B, and also with isolated LDL directly coated onto the microtiter plate. In Western blotting after SDS-PAGE Clone 02 binds to apo B in reduced as well as in non-reduced forms, using isolated LDL as antigen.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit