Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein C2 Human ELISA Kit Non-distribution product as customer accommodation.

Sandwich ELISA Kit

$485.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 1.5 ng/mL
Assay Range 1.56-100 ng/mL
Test Time 1 hr. 20 min.
Sample Volume Plasma, 2 μL; Serum, 2 μL; Supernatant, 100 μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHAPOC2 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
EHAPOC2
96 Tests
Pack for $485.00
Add to cart
 
Description

Description

The Human Apolipoprotein C2 (Hu ApoC2) ELISA quantitates Hu ApoC2 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoC2. Principle of the method The Human ApoC2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues .
Specifications

Specifications

2°C to 8°C
1.56-100 ng/mL
Plasma, 2 μL; Serum, 2 μL; Supernatant, 100 μL
<10%
1.5 ng/mL
1 hr. 20 min.
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit