Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein D Mouse anti-Human, Janelia Fluor 646, Clone: APOD/3415, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314090JF646
This item is not returnable.
View return policy
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Apolipoprotein D | |
Monoclonal | |
Janelia Fluor 646 | |
apo-D, ApoD, apolipoprotein D | |
Human recombinant Apolipoprotein D protein fragment (around aa21-119) (exact sequence is proprietary) (Uniprot: P05090) | |
0.1 mL | |
Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
347 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
APOD/3415 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title