Learn More
ApolipoproteinB/ApoB Mouse anti-Human, Janelia Fluor 646, Clone: 7B8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223519JF646
Catalog No. NBP23519646
Add to cart
Description
Description
ApolipoproteinB/ApoB Monoclonal antibody specifically detects ApolipoproteinB/ApoB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
ApolipoproteinB/ApoB
7B8
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
APOB
Human LDL
Protein A or G purified
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
338
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Mouse
IgG2b Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone 02 reacts with apo B from human serum/plasma, when used in sandwich ELISA in combination with a polyclonal antibody against apo B, and also with isolated LDL directly coated onto the microtiter plate. In Western blotting after SDS-PAGE Clone 02 binds to apo B in reduced as well as in non-reduced forms, using isolated LDL as antigen.