Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein M Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549077

Catalog No. PA549077

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.
Specifications

Specifications

Apolipoprotein M
Polyclonal
PBS with 0.09% sodium azide
Q9Z1R3
Apolipoprotein M, Apo-M, ApoM, Apom, Ng20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55938
Western Blot
Unconjugated
Apom
Liquid
APOM
KLH conjugated synthetic peptide between 93-119 amino acids from the Central region of mouse Apom.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit