Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apomorphine Hydrochloride, Hemihydrate, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
500 g
Packaging:
Amber Glass Bottle
41372-20-7
41372-20-7

A1147, 41372-20-7, C17H17NO2•HCl•1/2H2O

Spectrum™ Apomorphine Hydrochloride, Hemihydrate, USP is a non-selective dopamine agonist used in the treatment of Parkinson′s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN2811
Percent Purity 98.5 to 101.5%
Packaging Amber Glass Bottle
Quantity 500 mg