Learn More
MilliporeSigma™ ApopTag™ TdT Enzyme
Qualified for use with Apoptag In Situ Apoptosis Detection Kits
Supplier: MilliporeSigma™ S7107
Includes: Each vial contains 300μL of ApopTag Terminal Transferase (TdT) Enzyme
Description
A recombinant enzyme intended for use in labeling the 3'-OH ends of fragmented DNA during apoptotic cell detectionSpecifications
-15°C to -25°C | |
Labeling the 3'-OH ends of fragmented DNA during apoptotic cell detection | |
300 μL |