Learn More
Alfa Aesar™ Apoptosis Activator 2
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
Manufacturer:  Alfa Aesar™ J65320MB
 View more versions of this product
Catalog No. AAJ65320MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Orange
Powder
2° to 8°C
C15H9Cl2NO2
1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione
Apoptosis Reagent
25mg
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
79183-19-0
306.14g/mol
Light Sensitive