Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Antibody (OTI4C9) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270197

Catalog No. NBP270197


Add to Cart

Description

Description

APP Monoclonal specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

APP
Monoclonal
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Mouse
85.1 kDa
100 μg
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunohistochemistry
OTI4C9
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
APP
Full length human recombinant protein of human APP(NP_000475) produced in HEK293T cell.
Affinity Purified
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
351
Human, Mouse, Rat
Lyophilized
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit