Learn More
APP Antibody (OTI4C9), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270197UV
Catalog No. NBP270197UV
Add to Cart
Description
Description
APP Monoclonal specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
APP
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
APP
Full length recombinant protein of human APP (NP_000475) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Immunohistochemistry
OTI4C9
Western Blot, Immunohistochemistry
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Mouse
Affinity Purified
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
351.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions