Learn More
APP Human anti-Human, Alexa Fluor 647, Clone: Hu13, R&D Systems™
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9919R100UG
Catalog No. FAB9919R-100
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
APP Monoclonal antibody specifically detects APP in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
APP
Hu13
Flow Cytometry 0.25-1μg/106 cells
Supernatant
Human
IgG1
100μg
Primary
351
Human
Flow Cytometry
Alexa Fluor 647
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Human APP/Protease Nexin II
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects Human APP/Protease Nexin II based on Solanezumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Solanezumab. This product is for research use only.