Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APP Mouse anti-Human, Alexa Fluor™ 750, Clone: Hu15, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9710S100UG
This item is not returnable.
View return policy
Description
APP Monoclonal specifically detects APP in Human samples. It is validated for Flow Cytometry.Specifications
APP | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human APP/Protease Nexin II based on Aducanumab therapeutic antibody. This non-therapeutic uses the same variable region sequence as the therapeutic antibody Aducanumab. This product is for research use only. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
Hu15 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II | |
Human APP/Protease Nexin II | |
100 μg | |
351.0 | |
Human | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title