Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Mouse anti-Human, Clone: OTI1E6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504968

Catalog No. 50-167-5543


Add to cart

Description

Description

Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa.
Specifications

Specifications

APP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P05067
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2, preA4
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
351
Immunocytochemistry, Immunofluorescence, Western Blot
OTI1E6
Unconjugated
APP
Liquid
APP
Full length human recombit protein of human APP produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit