Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Mouse anti-Human, PE-Cyanine7, Clone: OTI7G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270187PECY7

Catalog No. N270187PEC7


Add to cart

Description

Description

APP Monoclonal antibody specifically detects APP in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

APP
OTI7G9
Flow Cytometry
Affinity Purified, Conjugated
APP
Full length recombinant protein of human APP (NP_000475) produced in HEK293T cells.
Immunogen affinity purified
Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary
351.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit