Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP (phospho Y757) Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30193

Catalog No. 89-033-731


Add to cart

Description

Description

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human APP.

  • High specificity
  • High reactivity
Specifications

Specifications

APP (phospho Y757)
Unconjugated
ELISA (0.05-0.2 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
P05067
AAA|ABETA|ABPP|AD1|APPI|CTFgamma|CVAP|PN2
Rabbit
IgG
40 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human APP.
In PBS, pH 7.4 (30% glycerol, 0.02% sodium azide).
Liquid
APP
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y757 of human APP.
Affinity purification
RUO
Primary
351
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)