Learn More
APP Rabbit anti-Human,Mouse,Rat, KHL, Clone: 5D16, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB115125UL
 View more versions of this product
Catalog No. ZRB115125MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
APP Monoclonal antibody specifically detects APP in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
APP
5D16
KHL
Purified
APP, A4, AD1
KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region.
Target molecular weight 86.94 kDa calculated; ∼100 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_958817.1
APP;ABPP;APPI;Alzheimer disease amyloid protein;Amyloid precursor protein;Amyloid-beta A4 protein;Cerebral vascular amyloid peptide;CVAP;PreA4;Protease nexin-II;PN-II
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human, Mouse, Rat