Learn More
APPBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148631AP
Catalog No. 50-172-9786
Add to cart
Description
Description
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer ft.s disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
APPBP1
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13564, Q8VBW6, Q9Z1A5
A 116A10.1, APP BP1, APPBP1, HPP1, NAE1, Proto oncogene protein 1, ula 1
Rabbit
IgG
150 μL
-20°C
Primary
234664, 84019, 8883
Immunoprecipitation, Western Blot
Unconjugated
NAE1
Liquid
NAE1
APPBP1 Fusion Protein Ag6672
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat