Learn More
APPL1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ARC1356, Invitrogen™
Rabbit Monoclonal Antibody
Manufacturer:  Invitrogen MA535737
Catalog No. PIMA535737
Add to cart
Description
Description
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
Specifications
Specifications
APPL1
ARC1356
PBS with 50% glycerol and 0.2% sodium azide; pH 7.3
Q8K3H0, Q9UKG1
2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; AI585782; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; Dip13a; DIP13alpha; Dip13-alpha; KIAA1428; MODY14; RGD1309388
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
26060, 290537, 72993
Western Blot
Unconjugated
APPL1
Liquid
APPL1
A synthesized peptide derived from human APPL
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat