Learn More
AmpliTaq™ 360 DNA Polymerase Non-distribution product as customer accommodation.
Amplifies vast range of DNA sequence contexts when used with enhanced AmpliTaq 360 Buffer and 360 GC Enhancer
Manufacturer:  Applied Biosystems 4398893
 View more versions of this product
Catalog No. 43-988-93
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
5' - 3'
High
3'-A
3000 units
When stored at -15 to -25 °C, this product maintains performance at least until the expiration date on the tube label. Before use, thaw frozen components to room temperature and mix well. Product is guaranteed up to 12 months as indicated by the expiration date.
1 X
AmpliTaq Gold 360 DNA Polymerase is supplied at 5U/μL with 10X AmpliTaq Gold 360 PCR Buffer, 25mM M2Cl2 , and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq Gold 360 Buffer, 25mM MgCl2 , and 360 GC Enhancer is supplied with each 250U of enzyme. Storage Buffer Enzyme is supplied in 20mM Tris, pH 9.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 0.5% (w/v) Tween 20, 50% (v/v) glycerol. Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.
AmpliTaq Gold™ 360 DNA Polymerase
5 kb or less