AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation
Manufacturer: Applied Biosystems 4311814
 View more versions of this product
Catalog No. 43-118-14
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR
    Includes 6 tubes (50μL at 5U/μL) of AmpliTaq Gold DNA Polymerase 6 tubes (1.5mL) of GeneAmp™ 10X Gold Buffer (150mM Tris-HCl, pH 8.0, 500mM KCl); 6 tubes (1.5mL) of 25mM magnesium chloride.
    Overhang 3'-A
    Quantity 1500 units
    Size (Final Product) 5kb or less
  • Description & Specifications
Related Products
Product Content Feedback