Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmpliTaq Gold™ DNA Polymerase, LD (Low DNA) with Gold Buffer and MgCl2 Non-distribution product as customer accommodation.

Purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations

Manufacturer:  Applied Biosystems 4338857

 View more versions of this product

Catalog No. 43-388-57


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications Description & Specifications

Specifications

5' - 3'
Low
3'-A
1000 units
Store at -15 to -30°C.
1 X
Contains AmpliTaq Gold& DNA Polymerase, LD (1000 units), GeneAmp& 10X PCR Gold Buffer, and MgCl2 Solution.
AmpliTaq Gold™ DNA Polymerase
5 kb or less
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit