Learn More
Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix
Premix, hot start chemistry for end-point PCR analysis
Supplier: Applied Biosystems™ 4390937
Includes: This master mix comes in a 250μL tube, enough for 25 reactions.
Description
Optimized chemical design shortens PCR running time from two hours to approximately 40 minutes yet generates high-quality PCR products that provide high-quality sequencing results. Optimized for use in sequencing applications, kit provides specific, high-yield amplicons that can be easily and robustly sequenced. This premixed master mix works with existing primers to provide sensitive, specific and reproducible results.
- Fast Results: Speeds time from sample to results
- Amplifies 600bp length gene in approximately 45 minutes
- Better Sequencing quality: Specific, high-yield amplicons suitable for sequencing applications that yield low peak-under-peak, longer continous read-length and higher QV-30 count
- Easy to use: The 2X master mix allows for quick experiment setup
- No primer annealing restrictions
See user's manual or package insert for limited label license, and trademark information
Applications:
Fast PCR, PCR, PCR and Real-Time PCR, Sanger Sequencing, Sanger Sequencing Technology and Accessories, Sequencing
Order Info
Shipping Condition: Wet ice
Specifications
This master mix comes in a 250μl tube, enough for 25 reactions. | |
Tube | |
AmpliTaq Gold DNA Polymerase | |
5' - 3' | |
Fast PCR Master Mix | |
Wet Ice | |
Cloning | |
1 X | |
25 Reactions | |
SuperMix or Master Mix | |
DNA |
Primer-probe | |
High | |
Fast | |
AmpliTaq Gold™ | |
25 Reactions | |
25 Reactions | |
2X | |
Built-In Hot Start | |
3'-A | |
1.5 kb or less |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.