Learn More
GeneAmp™ Fast PCR Master Mix (2X) Non-distribution product as customer accommodation.
Sensitive, pre-mixed hot-start chemistries
Manufacturer:  Applied Biosystems 4362070
 View more versions of this product
Catalog No. 43-620-70
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
5' - 3'
Medium
250.0
AmpliTaq™ DNA Polymerase
2 kb or less
1 X
The GeneAmp Fast PCR Master Mix contains a hot start polymerase system that does not require an activation step and has been optimized to decrease the overall PCR amplification time. The Fast PCR Master Mix is provided at 2X concentration for easier preparation of PCR reactions requiring only the addition of primers and templates. The GeneAmp Fast PCR Master Mix contains two tubes (1.25mL each) sufficient for 125 20μL PCR reactions. The Fast PCR Master Mix can also be ordered without a protocol.
3'-A
250 reactions, with protocol
Store at 2 to 8 °C.