GeneAmp™ High Fidelity PCR System Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Amplify DNA fragments up to 10kb
Manufacturer: Applied Biosystems 4328217
 View more versions of this product
Catalog No. 43-282-17
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Fidelity (vs. Taq) Fidelity (vs. Taq): 3X
    Includes Contains GeneAmp High Fidelity Enzyme Mix, 10X PCR Buffer with MgCl2, 10X PCR Buffer without MgCl2, and MgCl2 solution. Sufficient for 200 PCR amplifications, 50μL each.
    Quantity 200 reactions
    Storage Requirements Store at -20°C.
  • Description & Specifications
Related Products
Product Content Feedback