Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Multicolor Reaction Plates with Barcode
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Supplier: Applied Biosystems™ 4483317
Description
Applied Biosystems MicroAmp EnduraPlate microplates offer excellent PCR or qPCR performance in formats developed to meet experimental needs. All plates are validated with Applied Biosystems instruments for optimal fit and performance.
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
- Available in multiple colors for easy organization of laboratory assays
- Great mechanical stability and flatness for complete workflow needs
- Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and high durability
- Ideal choice for use with automation/robotics and multi-instrument experiments (like thermal cycler, real-time thermal cycler, genetic analyzer)
- Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
- Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity
- Platform: 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7900HT System, GeneAmp™ 9700, QuantStudio™, QuantStudio Dx, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
Includes Barcode | |
Store at room temperature. | |
MicroAmp | |
Microplate | |
Skirted | |
40 μL | |
500 plates |
Certified DNA/RNase Free | |
500 Plates | |
Optical, Assorted | |
Thermal Cycler, Genetic Analyzer, DNA Analyzer | |
Polycarbonate, Polypropylene | |
384 | |
Standard |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.