Learn More
Applied Biosystems™ Path-ID™ Multiplex One-Step Kit
The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR).
Supplier: Applied Biosystems™ 4428207
Includes: 1.375mL of 2X Multiplex RT-PCR Buffer, 280μL of 10X Multiplex Enzyme Mix including AmpliTaq Gold and ArrayScript, 1.75mL of Nuclease-free water
Description
It is optimized for the amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer/probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. The kit contains a Multiplex Enzyme Mix with ArrayScript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.
Multiplex One-Step Kit is optimized for amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer/probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. Kit contains a Multiplex Enzyme Mix with Arrayscript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.
Applications:
One-Step qRT-PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Order Info
Built-In Hot Start. Guaranteed minimum shelf life is 60 days.
Specifications
•7 mL of 2X Multiplex RT-PCR Buffer • 1.4 mL of 10X Multiplex Enzyme Mix including AmpliTaq Gold and ArrayScript • 25 mL of Nuclease-free water • Store the kit at -20°C; the Nuclease-free water may be stored at -20°C, 4°C, or room temperature Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Liquid | |
1-step RT-qPCR, Multiplex qPCR | |
Standard | |
ROX (Pre-mixed) | |
One-Step RT-PCR Kit | |
500 Reactions | |
Built-In Hot Start | |
Total RNA, Viral RNA, mRNA |
Primer-probe | |
Tube(s) | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
Path-ID™ | |
ArrayScript™ | |
Pathogen Detection, Virus Detection | |
500 Reactions |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.