Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Gene Expression Master Mix Non-distribution product as customer accommodation.

Precise and reliable real-time qPCR quantification over 9 logs of linear dynamic range with Applied Biosystems™ TaqMan™ Gene Expression Master Mix.

Manufacturer:  Applied Biosystems™ 4369514

 View more versions of this product

Catalog No. 43-695-14


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Requires : Template and primers

Specifications

Description

TaqMan™ Gene Expression Master Mix has been validated with TaqMan™ Gene Expression Assays and Applied Biosystems™ real-time systems, providing precise quantification for a variety of gene expression-based real-time qPCR applications, including rare transcript detection, gene expression analysis, pathogen detection and viral load quantification,co-amplification of two targets, as well as specific detection of gene family members
TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and Uracil-DNA Glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX™ dye for superb precision on Applied Biosystems™ real-time PCR instruments.

  • Optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target
  • Extended benchtop stability provides consistent results for automated liquid-handling systems, making it the choice for high-throughput sample processing, overnight or over the weekend
Description & Specifications

Specifications

ROX (Pre-mixed)
2 x 5mL
Pharma and biopharma, real time PCR (qPCR), real time PCR-based gene expression profiling, target and lead identification and validation
cDNA, DNA (Genomic)
Store at 2°C to 8°C.
Detection Method: Primer-Probe Detection, PCR Method: qPCR
TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components. 2 Bottles (5mL each) 2X TaqMan™ Gene Expression Master Mix sufficient for 400 reactions.
7300 System, StepOnePlus™, Standard Mode, 7000 System, 7900HT System, 7500 System, StepOne™, Standard Mode

Safety and Handling

Recommended Storage : Store at 2° to 8°C.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit