Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562388
Description
Immunogen sequence: QHVNCPGGIN AWNTITSYID NQICQGQKNL CNNTGDPEMC PENGSCVPDG PGLLQCVCAD GFHGYKCMRQ G Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
Specifications
APR3 | |
Polyclonal | |
Unconjugated | |
ATRAID | |
0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; all-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein APR-3; apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; C11H2orf28; C17H2orf28; C2orf28; H1E6 protein; HSPC013; LOW QUALITY PROTEIN: all-trans retinoic acid-induced differentiation factor; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51374 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UW56 | |
ATRAID | |
Recombinant protein corresponding to Human APR3. Recombinant protein control fragment (Product #RP-101318). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction