Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Apramycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Click to view available options
Quantity:
25 mL
50 mL
Description
Apramycin sulfate is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Apramycin sulfate is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Apramycin sulfate is a useful biochemical for antibiotic and antibacterial research to study antibiotic resistance through protein synthesis inhibition by blocking translocation.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Type | Apramycin Sulfate |
Physical Form | Liquid |
Solubility Information | It is soluble in water. |
Quantity | 25 mL |
Chemical Name or Material | Apramycin sulfate, 50mg/mL in distilled water, sterile-filtered |
Safety and Handling
Hazard Category | H360 |
Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 265-890-7 |
TSCA | No |
storageNote1 | -30° to -10°C |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction