Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APRIL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118591
Description
Antibody detects endogenous levels of total APRIL/TNFSF13.
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
Specifications
APRIL | |
Polyclonal | |
Unconjugated | |
TNFSF13 | |
2310026N09Rik; A proliferation-inducing ligand; APRIL; CD256; TALL2; TALL-2; TNF- and APOL-related leukocyte expressed ligand 2; TNF-related death ligand 1; Tnfsf13; TNLG7B; Trdl1; TRDL-1; tumor necrosis factor (ligand) superfamily, member 13; tumor necrosis factor ligand 7B; tumor necrosis factor ligand superfamily member 13; tumor necrosis factor superfamily member 13; tumor necrosis factor-like protein ZTNF2; tumor necrosis factor-related death ligand-1; UNQ383/PRO715; ZTNF2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
287437, 69583, 8741 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75888, Q9D777 | |
TNFSF13 | |
A synthesized peptide derived from human TNFSF13(Accession O75888), corresponding to amino acid residues Y163-S213. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction