Learn More
DuPont™ Tychem 6000FR Aprons
DuPont™ Tychem 6000 FR Sleeved Apron. Elastic Wrists. Two buckle closure system. Taped Seams.
Manufacturer:  DuPont™ TP275TGYLG000200
 View more versions of this product
Catalog No. 19-122-050
Add to cart
Description
Description
Tychem 6000 FR offers a single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc. Tychem 6000 FR garments are designed to combine the trusted chemical protection of Tychem and thermal/arc protection of Nomex into a single layer garment. Tychem 6000 FR garments are designed to help provide industrial workers and Hazmat responders with permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents and to provide escape time from flash fires and protection from electric arc. Tychem 6000 FR fabric provides at least eight hours breakthrough protection against 17 of the 21 standard ASTM F1001 chemicals and at least 30 minutes of protection for more than 180 chemical challenges including chemical warfare agents. The arc rating of Tychem 6000 FR fabric is 15 cal/cm Ebt. This exceeds the NFPA 70E Hazard Risk Category 2 requirement of 8 cal/cm. Typical Applications: HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories.Tychem Thermopro for triple hazard protectionProvides frontal protectionFor flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.Sewn with Nomex threadTwo buckle closure systemOpen back for wearer comfortIdeal for use on top of an existing flame resistant (FR) garment where extra chemical protection is requiredMade in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Specifications
Gray
Large