Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Aprotinin, Analytical Grade, MP Biomedicals™

Forms stable complexes with enzymes. Aprotinin, Analytical Grade, MP Biomedicals is a competitive serine proteinase inhibitor that works by blocking the active sites of enzymes. This binding is reversible with most aprotinin-protease complexes, dissociati

$382.00 - $1113.00

Specifications

Form Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP219038210
View Documents
MP Biomedicals™
0219038210
10 mg
Each for $382.00
Add to cart
 
MP219038250
View Documents
MP Biomedicals™
0219038250
50 mg
Each for $1,113.00
Add to cart
 
Description

Description

Aprotinin is found in bovine lymph nodes, lung, parotid gland, spleen, liver, pancreas, seminal vesicles, thyroid gland, kidney, mucous membranes of the trachea and esophagus, ovaries, heart, posterior pituitary and cartilage. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, kallkrein, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases. It does not inhibit thrombin. Uses include:

  • Small amounts can be added to tubes of drawn blood to enable laboratory measurement of rapidly degraded proteins, such as glucagon.
  • Can be labelled with fluorescein isothiocyanate for use as a fluorescent histochemical reagent for staining glycoconjugates (mucosubstances) that are rich in uronic or sialic acids. The conjugate retains its antiproteolytic and carbohydrate-binding properties.
  • Used in cell biology as an enzyme inhibitor to prevent protein degradation during lysis or homogenization of cells and tissues.
Specifications

Specifications

Powder
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit