Learn More
Thermo Scientific Chemicals Aprotinin, from Bovine Lung
Aprotinin (iniprol, CAS # 9087-70-1) is a single polypeptide chain from bovine tissues with antifibrinolytic activities.
Supplier: Thermo Scientific Chemicals J63039MB
Description
A potent protease inhibitor.Aprotinin acts as a reversible serine protease blocking and a binding agent. It is widely employed as an inhibitor of trypsin. It is also used as a proteolytic inhibitor in radioimmunoassays of polypeptide hormones. It is involved in the purification of urokinase, trypsin, and chymotrypsin on immobilized aprotinin. Further, it is used in the quantification of kallikrein activity in mixtures of esterases and proteases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Aprotinin is a 58-amino acid polypeptide chain found in lung tissue
• Aprotinin can suppress the systemic inflammatory response by reversibly inhibiting various esterases and proteases; it acts as a broad spectrum serine protease inhibitor
Application
• Aprotinin can be used as an enzyme inhibitor in laboratory studies on chymotrypsin, kallikrein, plasmin, and others
• Aprotinin is useful for the laboratory study of post-operative bleeding and wound healing
Specifications
Protease Inhibition | |
25mg | |
2° to 8°C | |
Aprotinin, from Bovine Lung | |
Lyophilized |
Lyophilized powder | |
Bovine lung | |
RUO | |
Native |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only